Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PNKD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552695
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552695 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552695 Supplier Invitrogen™ Supplier No. PA552695
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GATANKASHN RTRALQSHSS PEGKEEPEPL SPELEYIPRK RGKNPMKA Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

Probable hydrolase that plays an aggravative role in the development of cardiac hypertrophy via activation of the NF-kappa-B signaling pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PNKD
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PNKD
Gene Accession No. Q8N490
Gene Alias 2210013N15Rik; 2810403H05Rik; AI854243; brain protein 17; Brp17; DYT8; FKSG19; FPD1; hydrolase PNKD-like protein; KIAA1184; KIPP1184; LOC106780846; LOW QUALITY PROTEIN: probable hydrolase PNKD; MNCb-5687; MR1; MR-1; Myofibrillogenesis regulator 1; paroxysmal nonkinesigenic dyskinesia; paroxysmal nonkinesiogenic dyskinesia; paroxysmal nonkinesiogenic dyskinesia protein; PDC; PKND1; Pnkd; PNKD metallo-beta-lactamase domain containing; PNKD, MBL domain containing; probable hydrolase PNKD; probable hydrolase PNKD; LOW QUALITY PROTEIN: probable hydrolase PNKD; similar to brain protein 17 isoform 2; TAHCCP2; Trans-activated by hepatitis C virus core protein 2; UNQ2491/PRO5778
Gene Symbols PNKD
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PNKD. Recombinant protein control fragment (Product #RP-90216).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25953
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.