Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PNKD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 115841AP150UL

Description
Probable hydrolase that plays an aggravative role in the development of cardiac hypertrophy via activation of the NF-kappa-B signaling pathway.Specifications
PNKD | |
Polyclonal | |
Unconjugated | |
PNKD | |
BRP17, DYT8, FKSG19, FPD1, KIAA1184, KIPP1184, MR 1, MR1, Myofibrillogenesis regulator 1, PDC, PNKD, Probable hydrolase PNKD, TAHCCP2, UNQ2491/PRO5778 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100188944, 25953, 56695 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q69ZP3, Q8N490 | |
PNKD | |
PNKD Fusion Protein Ag2176 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction