Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PNLDC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562535
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562535 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562535 Supplier Invitrogen™ Supplier No. PA562535
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VGHNMMMDLL HLHEKFFRPL PESYDQFKQN IHSLFPVLID TKSVTKDIWK EMNFPRVSNL SEVYEVLNSD LNPT Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.

3'-exoribonuclease that has a preference for poly(A) tails of mRNAs, thereby efficiently degrading poly(A) tails (PubMed:27515512). Exonucleolytic degradation of the poly(A) tail is often the first step in the decay of eukaryotic mRNAs and is also used to silence certain maternal mRNAs translationally during oocyte maturation and early embryonic development (PubMed:27515512). May act as a regulator of multipotency in embryonic stem cells. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PNLDC1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PNLDC1
Gene Accession No. Q8NA58
Gene Alias HsPNLDC1; PARN like, ribonuclease domain containing 1; PARN-like domain-containing protein 1; PNLDC1; poly(A)-specific ribonuclease (PARN)-like domain containing 1; Poly(A)-specific ribonuclease domain-containing protein 1; poly(A)-specific ribonuclease PARN-like domain-containing protein 1; Poly(A)-specific ribonuclease PNLDC1
Gene Symbols PNLDC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PNLDC1. Recombinant protein control fragment (Product #RP-101348).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 154197
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.