Learn More
Promega pNLF1-C [CMV/Hygro] Vector
Catalog No. prn1361
Click to view available options
Quantity:
20 μg
Generate C-Terminal Fusions with NanoLuc™ Enzyme. The pNLF1 Vectors use traditional cloning with a multiple cloning site (MCS) to generate C-terminal fusions to NanoLuc™ luciferase. All vectors contain a mammalian selectable marker to create a stable line.
  • Generate N- or C-Terminal Fusions with NanoLuc(R) Enzyme or N-Terminal Secreted NanoLuc(R) Luciferase
  • Use traditional cloning to insert protein of interest into the multiple cloning site (MCS)
  • Bright NanoLuc(R) reporter enables endogenous expression levels of fusion proteins for biologically relevant results
  • Choose from transient transfection or select for stable expression using hygromycin
Specifications
Quantity 20 μg
Content And Storage -30°C to -10°C
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.