Learn More
Promega pNLF1-NRF2 [CMV/neo] Vector
Catalog No. prn1391
Click to view available options
Quantity:
1 Ea.
NanoLuc Stability Sensors are ready-to-use vector systems that utilize the advantages of the NanoLuc luciferase reporter for stability studies of two key signaling proteins, HIF1A and NRF2, providing a way to directly measure the primary signaling event.
  • Vectors to Measure Intracellular Protein Turnover of HIFA and NRF2
  • Ready-to-use constructs are predesigned, optimized and tested for low endotoxin levels
  • C-terminal fusion of HIF1A NanoLuc(R) luciferase reporter in pNLF1-HIF1A [CMV/neo] Vector
  • NRF2 vector system has a pKEAP1-expressing vector for regulating intracellular NRF2 levels and pNLF1-NRF2 [CMV/neo] Vector expressing a C-terminal fusion with NanoLuc(R) luciferase
Specifications
Quantity 1 Ea.
Content And Storage -30°C to -10°C
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.