Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PNLIP Recombinant Rabbit Monoclonal Antibody (HL1310)
GREENER_CHOICE

Catalog No. PIMA546996
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546996 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA546996 Supplier Invitrogen™ Supplier No. MA546996
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

This gene is a member of the lipase gene family. It encodes a carboxyl esterase that hydrolyzes insoluble, emulsified triglycerides, and is essential for the efficient digestion of dietary fats. This gene is expressed specifically in the pancreas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PNLIP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone HL1310
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene PNLIP
Gene Accession No. P16233, P27657, Q6P8U6
Gene Alias 1810007A24Rik; pancreatic lipase; pancreatic triacylglycerol lipase; pancreatic triglyceride lipase; PANLI; PL; PNLIP; PNLIPD; PNLIPRP2; PTL; RP11-33D13.1; triglyceride lipase
Gene Symbols PNLIP
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Pancreatic Lipase. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25702, 5406, 69060
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.