Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PNLIP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121002
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121002 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121002 Supplier Invitrogen™ Supplier No. PA5121002
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Mouse liver, Mouse pancreas. The target is usually found in the following locations: Secreted. Immunogen sequence: IGHSLGAHAA GEAGRRTNGT IGRITGLDPA EPCFQGTPEL VRLDPSDAKF VDVIHTDGAP IVPNLGFGMS QVVGHLDFFP NGGVEMPGCK KNILSQIVDI DGIWEGTRDF AACNHLRSYK YYTDSIVNPD GFAGFPCASY NVFTANKCFP CPSGGCPQMG HYADRYPGKT NDVGQKFYLD TGDASNFARW RYKVSVTLSG KKVTGHILVS LFGNKGNSKQ YEIFKGTLKP DSTHSNEFDS DVDVGDLQMV KFIWYNNVIN PTLPRVGASK IIVETNVGKQ FNFCSPETVR EEVLLTLTPC.

This gene is a member of the lipase gene family. It encodes a carboxyl esterase that hydrolyzes insoluble, emulsified triglycerides, and is essential for the efficient digestion of dietary fats. This gene is expressed specifically in the pancreas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PNLIP
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PNLIP
Gene Accession No. P16233, P27657, Q6P8U6
Gene Alias 1810007A24Rik; pancreatic lipase; pancreatic triacylglycerol lipase; pancreatic triglyceride lipase; PANLI; PL; PNLIP; PNLIPD; PNLIPRP2; PTL; RP11-33D13.1; triglyceride lipase
Gene Symbols PNLIP
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 166-465 of human PNLIP (NP_000927.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25702, 5406, 69060
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.