Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PNLIPRP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540797
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540797 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540797 Supplier Invitrogen™ Supplier No. PA540797
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: YADQFKGKTS AVEQTFFLNT GESGNFTSWR YKVSVTLSGK EKVNGYIRIA Sequence homology: Cow: 86%; Guinea Pig: 79%; Horse: 86%; Human: 100%; Mouse: 79%; Pig: 86%; Rabbit: 86%; Rat: 86%.

PNLIPRP2 is a lipase with broad substrate specificity. It can hydrolyze both phospholipids and galactolipids. It acts preferentially on monoglycerides, phospholipids and galactolipids. PNLIPRP2 also contributes to milk fat hydrolysis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PNLIPRP2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PNLIPRP2
Gene Accession No. P54317
Gene Alias Cytotoxic T-lymphocyte lipase; galactolipase; pancreatic lipase; pancreatic lipase related protein 2; pancreatic lipase related protein 2 (gene/pseudogene); pancreatic lipase-related protein 2; pancreatic lipase-related protein type 2; pancreatic lipase-related protein type 2 precursor; Plrp2; PL-RP2; PNLIPRP2; Secretory glycoprotein GP-3
Gene Symbols PNLIPRP2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human PNLIPRP2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5408
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.