Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PNLIPRP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598067
Description
PNLIPRP2 Polyclonal Antibody for Western Blot, IHC (P), ELISA
PNLIPRP2 is a lipase with broad substrate specificity. It can hydrolyze both phospholipids and galactolipids. It acts preferentially on monoglycerides, phospholipids and galactolipids. PNLIPRP2 also contributes to milk fat hydrolysis.
Specifications
PNLIPRP2 | |
Polyclonal | |
Unconjugated | |
PNLIPRP2 | |
Cytotoxic T-lymphocyte lipase; galactolipase; pancreatic lipase; pancreatic lipase related protein 2; pancreatic lipase related protein 2 (gene/pseudogene); pancreatic lipase-related protein 2; pancreatic lipase-related protein type 2; pancreatic lipase-related protein type 2 precursor; Plrp2; PL-RP2; PNLIPRP2; Secretory glycoprotein GP-3 | |
Rabbit | |
Protein G | |
RUO | |
5408 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
3 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
P54317 | |
PNLIPRP2 | |
Recombinant Human Pancreatic lipase-related protein 2 protein (305-460AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction