Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PNMA1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112658
Description
PNMA1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
PNMA1 encodes a protein that is highly restricted to the brain and testis. Anti-PNMA1 reacts mainly with subnuclear elements (including the nucleoli) and to a lesser degree the cytoplasm.
Specifications
| PNMA1 | |
| Polyclonal | |
| Unconjugated | |
| Pnma1 | |
| 37 kDa neuronal protein; 5730402C15Rik; MA1; Neuron- and testis-specific protein 1; paraneoplastic antigen MA1; Paraneoplastic antigen Ma1 homolog; paraneoplastic Ma antigen 1; paraneoplastic neuronal antigen MA1; paraneoplastic onconeuronal protein MA1; PNMA family member 1; PNMA1 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 70481, 9240 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 2 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8C1C8, Q8ND90 | |
| Pnma1 | |
| Recombinant Human Paraneoplastic antigen Ma1 protein (151-353AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction