Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PNMA3 Antibody (OTI1C3), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273485JF549
Description
PNMA3 Monoclonal specifically detects PNMA3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
PNMA3 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PNMA3 | |
Full length recombinant protein of human PNMA3 (NP_037496) produced in E.coli. | |
0.1 mL | |
29944.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI1C3 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
MA3MA5, MGC132756, MGC132758, paraneoplastic antigen MA3, paraneoplastic cancer-testis-brain antigen | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction