Learn More
PNMA3 Mouse anti-Human, Clone: OTI1C3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502336

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene belongs to the paraneoplastic antigen MA (PNMA) family, which shares homology with retroviral Gag proteins. The PNMA antigens are highly expressed in the brain and also in a range of tumors associated with serious neurological phenotypes. There are reports of the presence of a functional -1 ribosomal frameshift signal (consisting of a heptanucleotide shift motif followed 3′ by a pseudoknot structure) in this gene, however, the frame-shifted product has not been characterized. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
PNMA3 | |
Monoclonal | |
Unconjugated | |
PNMA3 | |
DmelCG13488 | |
Mouse | |
Affinity Chromatography | |
RUO | |
29944 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI1C3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9UL41 | |
PNMA3 | |
Full length human recombit protein of human PNMA3 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.