Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PNMA5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560796
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560796 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560796 Supplier Invitrogen™ Supplier No. PA560796
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QARSFSDSSP QTIQGGLPPL VKRRRLLGSE STRGEDHGQA TYPKAENQTP GREGPQAAGE ELGNEAGAGA MSHPK Highest antigen sequence identity to the following orthologs: Mouse - 39%, Rat - 45%.

This gene encodes a member of the paraneoplastic Ma antigen protein family. These proteins have been implicated in the development of paraneoplastic disorders resulting from an immune response directed against them. Paraneoplastic disorders are the result of an abnormal immune response to a tumor. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq, Oct 2011].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PNMA5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PNMA5
Gene Accession No. Q96PV4
Gene Alias KIAA1934; Paraneoplastic antigen-like protein 5; paraneoplastic Ma antigen family member 5; PNMA5; Tumor antigen BJ-HCC-25
Gene Symbols PNMA5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PNMA5. Recombinant protein control fragment (Product #RP-100471).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114824
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.