Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PNMA6A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | PNMA6A |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
PNMA6A Polyclonal specifically detects PNMA6A in Human samples. It is validated for Western Blot.Specifications
| PNMA6A | |
| Polyclonal | |
| Rabbit | |
| NP_116271 | |
| 84968 | |
| Synthetic peptide directed towards the N terminal of human PNMA6AThe immunogen for this antibody is PNMA6A. Peptide sequence FTVLGKVFREEDNATAALVELDREVNYALVPREIPGTGGPWNVVFVPRCS. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| MA6, MGC15827, paraneoplastic antigen like 6A, PNMA6 | |
| PNMA6A | |
| IgG | |
| 44 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title