Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PNMAL1 Polyclonal Antibody

PNMAL1 Polyclonal Antibody
Supplier: Thermo Scientific PA544706
Description
PNMAL1 Polyclonal Antibody for Western Blot
Specifications
| PNMAL1 | |
| Polyclonal | |
| Unconjugated | |
| PNMA8A | |
| Paraneoplastic antigen-like protein 8A; paraneoplastic Ma antigen family like 1; paraneoplastic Ma antigen family-like 1; PNMA8A; PNMAL1; PNMA-like protein 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 55228 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q86V59 | |
| PNMA8A | |
| Synthetic peptide directed towards the middle region of human PNMAL1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction