Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PNMAL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553018
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553018 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553018 Supplier Invitrogen™ Supplier No. PA553018
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KPKGINSNST ANLEDPEVGD AESMAISEPI KGSRKPCVNK EELALKKPMA KCAWKGPREP PQDARAEAES PGGASESDQD GGHESPPKKK AVAWVSAKNP APMRKKKKVS LGPVSYVLVD SEDGRKKPVM P Highest antigen sequence identity to the following orthologs: Mouse - 47%, Rat - 50%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PNMAL1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PNMA8A
Gene Accession No. Q86V59
Gene Alias Paraneoplastic antigen-like protein 8A; paraneoplastic Ma antigen family like 1; paraneoplastic Ma antigen family-like 1; PNMA8A; PNMAL1; PNMA-like protein 1
Gene Symbols PNMA8A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PNMAL1. Recombinant protein control fragment (Product #RP-89740).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55228
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.