Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PNMT Antibody (SR1514), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322526100UL
Description
PNMT Monoclonal antibody specifically detects PNMT in Human, Mouse samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
PNMT | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunofluorescence | |
SR1514 | |
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
EC 2.1.1, EC 2.1.1.28, Noradrenaline N-methyltransferase, PENTMGC34570, phenylethanolamine N-methylase, phenylethanolamine N-methyltransferase, PNMTase | |
A synthesized peptide derived from human PNMT (Uniprot #: P11086) | |
100 μg | |
Cancer | |
5409 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction