Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PNO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110188
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110188 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110188 Supplier Invitrogen™ Supplier No. PA5110188
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MESEMETQSA RAEEGFTQVT RKGGRRAKKR QAEQLSAAGE GGDAGRMDTE EARPAKRPVF PPLCGDGLLS GKEETRKIPV PANRYTPLKE NWMKIFTPIV EHLGLQIRFN LKSRNVEIRT CKETKDVSAL TKAADFVKAF ILGFQVEDAL ALIRLDDLFL ESFEITDVKP LKGDHLSRAI GRIAGKGGKT KFTIENVTRT RIVLADVKVH ILGSFQNIKM ARTAICNLIL GNPPSKVYGN IRAVASRSAD RF.

Positively regulates dimethylation of two adjacent adenosines in the loop of a conserved hairpin near the 3'-end of 18S rRNA (PubMed:25851604). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PNO1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PNO1
Gene Accession No. Q6VBQ8, Q9NRX1
Gene Alias 1810003N24Rik; AA407421; AU015468; AU019495; AU042323; Emi3; expressed during mesenchymal induction 3; Imi3; induced during mesenchymal induction 3; KHRBP1; KH-type RNA-binding protein 1; partner of NOB1 homolog; partner of NOB1 homolog (S. cerevisiae); Pno1; putatative 28 kDa protein; putative 28 kDa protein; RNA binding protein; RNA-binding protein PNO1; RRP20
Gene Symbols PNO1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-252 of human PNO1 (NP_064528.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 289809, 56902
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.