Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PNO1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | PNO1 |
---|---|
Concentration | 0.14 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-3233
![]() ![]() |
Proteintech Group Inc
210591AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Positively regulates dimethylation of two adjacent adenosines in the loop of a conserved hairpin near the 3′-end of 18S rRNA.Specifications
PNO1 | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q6VBQ8, Q9CPS7, Q9NRX1 | |
289809, 56902, 66249 | |
PNO1 Fusion Protein Ag14752 | |
Primary | |
-20°C | |
PNO1 |
0.14 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
KHRBP1, PNO1, RNA binding protein PNO1 | |
Pno1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title