Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PNOC Monoclonal Antibody (2133CT1268.37.7)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA537785
Description
PNOC Monoclonal Antibody for Western Blot
PNOC is the ligand of the opioid receptor-like receptor (OPRL1). It may act as a transmitter in the brain by modulating nociceptive and locomotor behavior. This protein may be involved in neuronal differentiation and development.
Specifications
PNOC | |
Monoclonal | |
Unconjugated | |
PNOC | |
N/OFQ; N23K; N23K/N27K; neuropeptide nociceptin 1; nociceptin; nociceptin/orphanin FQ precursor; nociceptin; prepronociceptin; nocistatin; NOP; Npnc1; OFQ; OFQ/N; ORL1 receptor agonist; Orphanin FQ; Orphanin FQ2; Pnoc; ppN/OFQ; PPNOC; pre-pro-N/OFQ; prepronociceptin; Prepronociceptin (neuropeptide nociceptin) (N23K); pronociceptin; proorphanin; propronociceptin | |
Mouse | |
Protein G | |
RUO | |
5368 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
2133CT1268.37.7 | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q13519 | |
PNOC | |
recombinant protein from human PNOC. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction