Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PNP Recombinant Rabbit Monoclonal Antibody (001)
GREENER_CHOICE

Catalog No. PIMA529490
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529490 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529490 Supplier Invitrogen™ Supplier No. MA529490
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human PNP.

The PNP gene encodes for the enzyme purine nucleoside phosphorylase, which plays a vital role in the purine salvage pathway by catalyzing the reversible cleavage of purine nucleosides. This enzyme facilitates the conversion of inosine, guanosine, and deoxyguanosine into free bases and sugars phosphorylated at the C1 position. PNP is located on chromosome 14q13 and is crucial for nucleoside catabolism, impacting energy metabolism and cellular functions across various organisms. PNP deficiency is an autosomal recessive multisystem disorder arising from mutations in the PNP gene, leading to impaired T-cell maturation and function, resulting in recurrent infections, autoimmune manifestations, and neurological abnormalities due to the toxic accumulation of deoxyguanosine triphosphate (dGTP). This deficiency particularly affects immature thymocytes and can lead to lymphoid and neuronal toxicity, underscoring the gene's importance in proper immune system function and cellular health.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PNP
Applications Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone 001
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene PNP
Gene Accession No. P00491
Gene Alias AL024301; AU015798; endothelial dysfunction inducing protein; epididymis secretory sperm binding protein Li 156an; HEL-S-156an; inosine phosphorylase; inosine phosphorylase {ECO:0000250; Inosine-guanosine phosphorylase; Np; Np-1; Np-2; nucleoside phosphorylase; PNP; PNP {ECO:0000250; Pnp1; PRO1837; PUNP; purine nucleoside phosphorylase; purine nucleoside phosphorylase {ECO:0000250; purine-nucleoside phosphorylase; purine-nucleoside phosphorylase 1; purine-nucleoside:orthophosphate ribosyltransferase; UniProtKB:P00491}
Gene Symbols PNP
Host Species Rabbit
Immunogen Recombinant Human PNP protein (Met1-Ser289).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4860
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.