Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PNPase Monoclonal Antibody (4C11)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA527859
Description
Positive Control: Jurkat, Raji, K562, THP-1 Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PNPT1 (polyribonucleotide nucleotidyltransferase 1, mitochondirla) is a RNA-binding protein implicated in numerous RNA metabolic processes. It catalyzes the phosphorolysis of single-stranded polyribonucleotides processively in the 3'-to-5' direction. PNPT1 is a component of the mitochondrial degradosome (mtEXO) complex that degrades 3' overhang double-stranded RNA with a 3'-to-5' directionality in an ATP-dependent manner. It is required for correct processing and polyadenylation of mitochondrial mRNAs. It plays a role as a cytoplasmic RNA import factor, in mitochondrial morphogenesis and respiration, regulation of expression of the electron transport chain, regulation of the stability of specific mature miRNAs in melanoma cells, and RNA cell surveilance. Mutations of the gene can result in combined oxidative phosphorylation deficiency 13 (COXPD13) and deafness, autosomal recessive, 70 (DFNB70).
Specifications
PNPase | |
Monoclonal | |
1 mg/mL | |
PBS with 20% glycerol and no preservative; pH 7 | |
Q8TCS8 | |
PNPT1 | |
Human PNPase. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunoprecipitation, Western Blot | |
4C11 | |
Unconjugated | |
PNPT1 | |
1200003F12Rik; 3'-5' RNA exonuclease OLD35; COXPD13; DFNB70; DKFZp762K1914; old 35; Old35; old-35; PNPase; PNPase 1; PNPase old-35; Pnpt1; Pnptl1; polynucleotide phosphorylase 1; Polynucleotide phosphorylase-like protein; polyribonucleotide nucleotidyltransferase 1; polyribonucleotide nucleotidyltransferase 1, mitochondrial | |
Mouse | |
Protein G | |
RUO | |
87178 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction