Learn More
PNPase Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303917AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse
PNPT1 is a subunit of the exosome complex, which is involved in 3-prime-to-5-prime exoribonuclease activity for RNA processing and degradation.PNPT1 is a subunit of the exosome complex, which is involved in 3-prime-to-5-prime exoribonuclease activity for RNA processing and degradation (Raijmakers et al., 2002 [PubMed 12419256]).[supplied by OMIM]. Sequence Note: This RefSeq record was created from transcript and genomic sequence data because no single transcript was available for the full length of the gene. The extent of this transcript is supported by transcript alignments. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.Specifications
PNPase | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q8TCS8 | |
PNPT1 | |
Between 700 and 750 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
PNPT1 | |
OLD35, PNPASE, old-35, 3′-5′ RNA exonuclease OLD35, PNPase 1, PNPase old-35, polynucleotide phosphorylase 1, polynucleotide phosphorylase-like protein, polyribonucleotide nucleotidyltransferase 1, mitochondrial | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
87178 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.