Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PNPLA1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | PNPLA1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
PNPLA1 Polyclonal specifically detects PNPLA1 in Human samples. It is validated for Western Blot.Specifications
PNPLA1 | |
Polyclonal | |
Rabbit | |
Lipid and Metabolism | |
Q8N8W4-2 | |
285848 | |
Synthetic peptides corresponding to PNPLA1 (patatin-like phospholipase domain containing 1) The peptide sequence was selected from the middle region of PNPLA1)(50ug). Peptide sequence QAPTSHRPSLGPSTVGAPQTLPRSSLSAFPAQPPVEELGQEQPQAVALLV. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
dJ50J22.1, EC 3.1.1.-, FLJ38755, MGC119319, MGC119320, MGC119321, patatin-like phospholipase domain containing 1, patatin-like phospholipase domain-containing protein 1 | |
PNPLA1 | |
IgG | |
48 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title