Learn More
Invitrogen™ PNPLA1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Supplier:  Invitrogen™ PA5113223
Catalog No. PIPA5113223
Only null left
Add to Cart
Description
Description
PNPLA1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Human patatin-like phospholipases, such as PNPLA1, have been implicated in regulation of adipocyte differentiation and have been induced by metabolic stimuli (Wilson et al., 2006 [PubMed 16799181]).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PNPLA1
Polyclonal
Unconjugated
PNPLA1
ARCI10; dJ50J22.1; patatin like phospholipase domain containing 1; patatin-like phospholipase domain containing 1; patatin-like phospholipase domain-containing protein 1; Pnpla1
Rabbit
Protein G
RUO
285848, 433091
-20°C or -80°C if preferred
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Q3V1D5, Q8N8W4
PNPLA1
Recombinant Human Patatin-like phospholipase domain-containing protein 1 protein (101-300AA).
100 μg
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.