Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PNPLA1 Polyclonal Antibody, Biotin, Bioss
SDP

Catalog No. 502281144
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-1144 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-1144 Supplier Bioss Supplier No. BS24622RBIOTIN
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PNPLA1 Polyclonal antibody specifically detects PNPLA1 in Rat samples. It is validated for Western Blot

Specifications

Antigen PNPLA1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Biotin
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene PNPLA1
Gene Alias ARCI10; dJ50J22.1; patatin like phospholipase domain containing 1; patatin-like phospholipase domain containing 1; patatin-like phospholipase domain-containing protein 1; Pnpla1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PNPLA1
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361812
Target Species Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.