Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PNPLA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543293
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543293 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543293 Supplier Invitrogen™ Supplier No. PA543293
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: QAPTSHRPSL GPSTVGAPQT LPRSSLSAFP AQPPVEELGQ EQPQAVALLV Sequence homology: Human: 93%.

Human patatin-like phospholipases, such as PNPLA1, have been implicated in regulation of adipocyte differentiation and have been induced by metabolic stimuli (Wilson et al., 2006 [PubMed 16799181]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PNPLA1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PNPLA1
Gene Accession No. Q8N8W4
Gene Alias ARCI10; dJ50J22.1; patatin like phospholipase domain containing 1; patatin-like phospholipase domain containing 1; patatin-like phospholipase domain-containing protein 1; Pnpla1
Gene Symbols PNPLA1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human PNPLA1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285848
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.