Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PNPLA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543256
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543256 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543256 Supplier Invitrogen™ Supplier No. PA543256
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: CSPKGCPAET KAEATPRSIL RSSLNFFLGN KVPAGAEGLS TFPSFSLEKS Sequence homology: Human: 100%.

The protein encoded by this gene is a triacylglycerol lipase that mediates triacylglycerol hydrolysis in adipocytes. The encoded protein, which appears to be membrane bound, may be involved in the balance of energy usage/storage in adipocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PNPLA3
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PNPLA3
Gene Accession No. Q9NST1
Gene Alias 1-acylglycerol-3-phosphate O-acyltransferase Pnpla3; acylglycerol O-acyltransferase; Acylglycerol transacylase; Adiponutrin; ADPN; C22orf20; Calcium-independent phospholipase A2-epsilon; iPLA(2)epsilon; iPLA2epsilon; iPLA2-epsilon; Lysophosphatidic acid acyltransferase; patatin like phospholipase domain containing 3; patatin-like phospholipase domain containing 3; patatin-like phospholipase domain-containing protein 3; Pnpla3
Gene Symbols PNPLA3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human PNPLA3 (aa 431-480).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80339
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.