Learn More
Invitrogen™ PNPLA3 Polyclonal Antibody

Description
In direct ELISAs, approximately 15% cross-reactivity with recombinant mouse Adiponutrin is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | PNPLA3 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | PNPLA3 |
| Gene Accession No. | Q9NST1 |
| Gene Alias | 1-acylglycerol-3-phosphate O-acyltransferase Pnpla3; acylglycerol O-acyltransferase; Acylglycerol transacylase; Adiponutrin; ADPN; C22orf20; Calcium-independent phospholipase A2-epsilon; iPLA(2)epsilon; iPLA2epsilon; iPLA2-epsilon; Lysophosphatidic acid acyltransferase; patatin like phospholipase domain containing 3; patatin-like phospholipase domain containing 3; patatin-like phospholipase domain-containing protein 3; Pnpla3 |
| Gene Symbols | PNPLA3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.