Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PNPLA5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP170680
Description
PNPLA5 Polyclonal specifically detects PNPLA5 in Human samples. It is validated for Western Blot.Specifications
| PNPLA5 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| PNPLA5 | |
| Synthetic peptides corresponding to PNPLA5(patatin-like phospholipase domain containing 5) The peptide sequence was selected from the C terminal of PNPLA5. Peptide sequence NMALEVFSRTKAQLLGPISPPATRVLETSPLQPQIAPHREELGPTHQA. | |
| Affinity purified | |
| RUO | |
| 150379 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| dJ388M5, dJ388M5.4, GS2L, patatin-like phospholipase domain containing 5, patatin-like phospholipase domain-containing protein 5 | |
| Rabbit | |
| 48 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Human: 100%. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction