Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PNPLA6 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$343.50 - $573.00

Specifications

Antigen PNPLA6
Dilution Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB169661
SDP
View Documents
Novus Biologicals
NBP31729225UL
25 μg
Each for $343.50
Only null left
Add to Cart
 
NB169660
SDP
View Documents
Novus Biologicals
NBP317292100UL
100 μg
Each for $573.00
Only null left
Add to Cart
 
Description

Description

PNPLA6 Polyclonal antibody specifically detects PNPLA6 in Human samples. It is validated for Immunofluorescence
Specifications

Specifications

PNPLA6
Immunofluorescence
Unconjugated
Rabbit
Human
EC 3.1.1.5, neuropathy target esterase, NTENTEMND, patatin-like phospholipase domain containing 6, Patatin-like phospholipase domain-containing protein 6, SPG39, sws
This antibody was developed against Recombinant Protein corresponding to amino acids: SYCEDESATGGCPFGPYQGRQTSSIFEAAKQELAKLMRIEDPSLLNSRVLLHHAKAGTIIARQGDQDVSLHFVLWG
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL
Polyclonal
Purified
RUO
PBS, pH 7.2, 40% glycerol
10908
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.