Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PNPLA6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | PNPLA6 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
PNPLA6 Polyclonal specifically detects PNPLA6 in Mouse samples. It is validated for Western Blot.Specifications
PNPLA6 | |
Polyclonal | |
Rabbit | |
Q3TRM4-3 | |
10908 | |
Synthetic peptides corresponding to the C terminal of Pnpla6. Immunizing peptide sequence DGCADGEESDCLTEYEEDAGPDCSRDEGGSPEGASPSTASEVEEEKSTLR. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
EC 3.1.1.5, neuropathy target esterase, NTENTEMND, patatin-like phospholipase domain containing 6, Patatin-like phospholipase domain-containing protein 6, SPG39, sws | |
PNPLA6 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title