Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PNPLA8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550992
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550992 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550992 Supplier Invitrogen™ Supplier No. PA550992
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PNPLA8 Polyclonal Antibody detects endogenous levels of PNPLA8 protein.

Phospholipase A2 catalyzes cleavage of fatty acids from phospholipids, thereby regulating membrane physical properties and the release of lipid second messengers and growth factors. Phospholipase A2 activity also modulates cellular growth programs, inflammation, and ion channel function (Mancuso et al., 2000 [PubMed 10744668]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PNPLA8
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PNPLA8
Gene Accession No. Q8K1N1, Q9NP80
Gene Alias 1200006O19Rik; AI467579; BM-043; Calcium-independent phospholipase A2-gamma; intracellular membrane-associated calcium-independent phospholipase A2 gamma; iPLA2 gamma; Ipla2(gamma); IPLA22; IPLA2-2; IPLA2G; iPLA2gamma; iPLA2-gamma; membrane-associated calcium-independent phospholipase A2 gamma; MMLA; patatin like phospholipase domain containing 8; patatin-like phospholipase domain containing 8; patatin-like phospholipase domain-containing protein 8; Pnpla8; PNPLA-gamma
Gene Symbols PNPLA8
Host Species Rabbit
Immunogen Synthesized peptide derived from the C-terminal region of human iPLA2G.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50640, 67452
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.