Learn More
PNPLA8 Polyclonal Antibody, Invitrogen™
Description
The antibody detects endogenous levels of total PNPLA8 protein.
Specifications
Specifications
| Antigen | PNPLA8 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | PNPLA8 |
| Gene Accession No. | Q8K1N1, Q9NP80 |
| Gene Alias | 1200006O19Rik; AI467579; BM-043; Calcium-independent phospholipase A2-gamma; intracellular membrane-associated calcium-independent phospholipase A2 gamma; iPLA2 gamma; Ipla2(gamma); IPLA22; IPLA2-2; IPLA2G; iPLA2gamma; iPLA2-gamma; membrane-associated calcium-independent phospholipase A2 gamma; MMLA; patatin like phospholipase domain containing 8; patatin-like phospholipase domain containing 8; patatin-like phospholipase domain-containing protein 8; Pnpla8; PNPLA-gamma |
| Gene Symbols | PNPLA8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.