Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PNPO Antibody (2C7), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197941H
Description
PNPO Monoclonal specifically detects PNPO in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PNPO | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
PNPO | |
Recombinant human PNPO (57-261aa) purified from E. coli | |
100 μL | |
Cancer, metabolism | |
55163 | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
2C7 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
EC 1.4.3.5, FLJ10535, PDXPO, pyridoxal 5'-phosphate synthase, pyridoxamine 5'-phosphate oxidase, Pyridoxamine-phosphate oxidase, pyridoxine 5'-phosphate oxidase, pyridoxine-5'-phosphate oxidase | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Anti-human PNPO mAb, clone 2C7, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human PNPO protein. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction