Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PNPO Antibody (OTI1H9), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273508V
Description
PNPO Monoclonal specifically detects PNPO in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
PNPO | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PNPO | |
Full length recombinant protein of human PNPO (NP_060599) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Western Blot, Immunohistochemistry | |
OTI1H9 | |
Western Blot, Immunohistochemistry | |
EC 1.4.3.5, FLJ10535, PDXPO, pyridoxal 5′-phosphate synthase, pyridoxamine 5′-phosphate oxidase, Pyridoxamine-phosphate oxidase, pyridoxine 5′-phosphate oxidase, pyridoxine-5′-phosphate oxidase | |
Mouse | |
Affinity Purified | |
Cancer, metabolism | |
55163.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction