Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PNPO Monoclonal Antibody (OTI3B3), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA503249S


Description
PNPO Monoclonal antibody specifically detects PNPO in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
PNPO | |
Monoclonal | |
0.57 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O75688 | |
Mouse | |
Affinity chromatography | |
RUO | |
5495 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2a |
Immunocytochemistry, Western Blot | |
OTI3B3 | |
Unconjugated | |
PPM1B | |
AI415282; epididymis secretory protein Li 302; FLJ10535; HEL-S-302; PDXPO; Pnpo; pyridoxal 5'-phosphate synthase; pyridoxamine 5'-phosphate oxidase; pyridoxamine-phosphate oxidase; pyridoxine 5'-phosphate oxidase; pyridoxine 5-phosphate oxidase; pyridoxine-5'-phosphate oxidase | |
Full length human recombinant protein of human PPM1B produced in HEK293 cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction