Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PNPO Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544902
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544902 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544902 Supplier Invitrogen™ Supplier No. PA544902
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PMRKSYRGDR EAFEETHLTS LDPVKQFAAW FEEAVQCPDI GEANAMCLAT Sequence homology: Cow: 86%; Dog: 93%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.

The enzyme encoded by this gene catalyzes the terminal, rate-limiting step in the synthesis of pyridoxal 5'-phosphate, also known as vitamin B6. Vitamin B6 is a required co-factor for enzymes involved in both homocysteine metabolism and synthesis of neurotransmitters such as catecholamine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PNPO
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PNPO
Gene Accession No. Q9NVS9
Gene Alias AI415282; epididymis secretory protein Li 302; FLJ10535; HEL-S-302; PDXPO; Pnpo; pyridoxal 5'-phosphate synthase; pyridoxamine 5'-phosphate oxidase; pyridoxamine-phosphate oxidase; pyridoxine 5'-phosphate oxidase; pyridoxine 5-phosphate oxidase; pyridoxine-5'-phosphate oxidase
Gene Symbols PNPO
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human PNPO (aa 36-85).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55163
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.