Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PNPO Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554722
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554722 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554722 Supplier Invitrogen™ Supplier No. PA554722
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSVIPDREYL RKKNEELEQL YQDQEVPKPK SWGGYVLYPQ VMEFWQGQTN RLHDRIVFRR GLPTGDSPLG PMTHRGEEDW LYERLAP Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 90%.

The enzyme encoded by this gene catalyzes the terminal, rate-limiting step in the synthesis of pyridoxal 5'-phosphate, also known as vitamin B6. Vitamin B6 is a required co-factor for enzymes involved in both homocysteine metabolism and synthesis of neurotransmitters such as catecholamine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PNPO
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PNPO
Gene Accession No. Q9NVS9
Gene Alias AI415282; epididymis secretory protein Li 302; FLJ10535; HEL-S-302; PDXPO; Pnpo; pyridoxal 5'-phosphate synthase; pyridoxamine 5'-phosphate oxidase; pyridoxamine-phosphate oxidase; pyridoxine 5'-phosphate oxidase; pyridoxine 5-phosphate oxidase; pyridoxine-5'-phosphate oxidase
Gene Symbols PNPO
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PNPO. Recombinant protein control fragment (Product #RP-93193).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55163
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.