Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PNPO Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986334
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6334 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6334 Supplier Bioss Supplier No. BS5108R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The enzyme encoded by this gene catalyzes the terminal, rate-limiting step in the synthesis of pyridoxal 5′-phosphate, also known as vitamin B6. Vitamin B6 is a required co-factor for enzymes involved in both homocysteine metabolism and synthesis of neurotransmitters such as catecholamine.

Specifications

Antigen PNPO
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PNPO
Gene Accession No. Q9NVS9
Gene Alias AI415282; BOS_18968; epididymis secretory protein Li 302; FLJ10535; HEL-S-302; LOC100349278; PDXPO; PNPO; pyridoxal 5′-phosphate synthase; pyridoxamine 5′-phosphate oxidase; Pyridoxamine-phosphate oxidase; pyridoxine 5′-phosphate oxidase; pyridoxine 5-phosphate oxidase; Pyridoxine-5′-phosphate oxidase
Gene Symbols PNPO
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PNPO.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55163
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.