Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PNPT1 Antibody (4C11), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP243725
Description
Ensure accurate, reproducible results in Western Blot, Immunoprecipitation
PNPT1 Monoclonal specifically detects PNPT1 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
PNPT1 | |
Monoclonal | |
Unconjugated | |
PBS (pH7), 20% Glycerol with No Preservative | |
PNPT1 | |
The immunogen used to generate this antibody corresponds to human PNPase. | |
100 μL | |
DNA replication Transcription Translation and Splicing | |
87178 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunoprecipitation | |
4C11 | |
Western Blot 1:500-1:3000, Immunoprecipitation | |
DKFZp762K1914, EC 2.7.7, EC 2.7.7.8, mitochondrial, old-35, OLD35, PNPase, PNPase 1, PNPase old-35, Polynucleotide phosphorylase 1, Polynucleotide phosphorylase-like protein, polyribonucleotide nucleotidyltransferase 1 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction