Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PNPT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | PNPT1 |
|---|---|
| Concentration | 0.37 mg/mL |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-9485
![]()
|
Proteintech Group Inc
144871AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody detected the endogenous PNPT1 around 80 kDa in mouse brain.
PNPase is involved with the exosome complex, which is involved in 3-prime-to-5-prime exoribonuclease activity for RNA processing and degradation.PNPT1 is a subunit of the exosome complex, which is involved in 3-prime-to-5-prime exoribonuclease activity for RNA processing and degradation.Specifications
| PNPT1 | |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q8K1R3, Q8TCS8 | |
| 360992, 71701, 87178 | |
| PNPT1 Fusion Protein Ag5888 | |
| Primary | |
| -20°C | |
| PNPT1 |
| 0.37 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| 3 5 RNA exonuclease OLD35, old 35, OLD35, PNPASE, PNPase 1, PNPase old 35, PNPT1, Polynucleotide phosphorylase 1 | |
| Pnpt1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title