Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PNPT1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987729
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7729 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7729 Supplier Bioss Supplier No. BS7100R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PNPT1 (polyribonucleotide nucleotidyltransferase 1, mitochondirla) is a RNA-binding protein implicated in numerous RNA metabolic processes. It catalyzes the phosphorolysis of single-stranded polyribonucleotides processively in the 3′-to-5′ direction. PNPT1 is a component of the mitochondrial degradosome (mtEXO) complex that degrades 3′ overhang double-stranded RNA with a 3′-to-5′ directionality in an ATP-dependent manner. It is required for correct processing and polyadenylation of mitochondrial mRNAs. It plays a role as a cytoplasmic RNA import factor, in mitochondrial morphogenesis and respiration, regulation of expression of the electron transport chain, regulation of the stability of specific mature miRNAs in melanoma cells, and RNA cell surveilance. Mutations of the gene can result in combined oxidative phosphorylation deficiency 13 (COXPD13) and deafness, autosomal recessive, 70 (DFNB70).

Specifications

Antigen PNPT1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PNPT1
Gene Accession No. Q8TCS8
Gene Alias 1200003F12Rik; 3′-5′ RNA exonuclease OLD35; BOS_11603; COXPD13; DFNB70; DKFZp762K1914; old 35; Old35; old-35; PNPASE; PNPase 1; PNPase old-35; Pnpt1; Pnptl1; Polynucleotide phosphorylase 1; Polynucleotide phosphorylase-like protein; polyribonucleotide nucleotidyltransferase 1; Polyribonucleotide nucleotidyltransferase 1, mitochondrial
Gene Symbols PNPT1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PNPT1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 87178
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.