Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PNRC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566848
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566848 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566848 Supplier Invitrogen™ Supplier No. PA566848
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PPLPRIPDPRA LPPTLFLPHF LGGDGPCLTP QPRAPAALPN RSLAVAGGTP RA Highest antigen sequence identity to the following orthologs - mouse 68%, rat 53%.

Nuclear receptor coactivator. May play a role in signal transduction. Classified as a a proline rich protein found enriched in bronchiolar epithelial cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PNRC1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PNRC1
Gene Accession No. Q12796
Gene Alias B4-2; PNAS-145; PNRC1; PROL2; proline rich 2 protein; proline rich nuclear receptor coactivator 1; proline-rich nuclear receptor coactivator 1; proline-rich polypeptide 2; proline-rich protein 2; proline-rich protein with nuclear targeting signal; protein B4-2; PRR2
Gene Symbols PNRC1
Host Species Rabbit
Immunogen Recombinant Human PNRC1. Recombinant protein control fragment (Product #RP-105395).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10957
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.