Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PNU 120596
GREENER_CHOICESDP

Catalog No. 24985 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-985 5 mg
24-982-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 24-985 Supplier Tocris Bioscience™ Supplier No. 2498/5
Add to Cart
Add to Cart
This item is not returnable. View return policy

Positive allosteric modulator of α7 nAChRs; active in vivo

PNU 120596 is a positive allosteric modulator of α7 neuronal nicotinic acetylcholine receptors (EC50 = 216 nM), with no detectable effect on α4β2, α3β4 and α9α10 receptors. Active in vivo following systemic administration. Neuroprotective in an in vivo model of transient focal cerebral ischemia.

Specifications

Chemical Name or Material N-(5-Chloro-2,4-dimethoxyphenyl)-N'-(5-methyl-3-isoxazolyl)-urea
CAS 501925-31-1
Assay Percent Range >99%
Quantity 5 mg
Molecular Formula C13H14ClN3O4
Formula Weight Observed MW: 311.72
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.