Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PNU 282987
GSA_VAGREENER_CHOICE

Catalog No. 230310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-031-0 10 mg
23-035-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-031-0 Supplier Tocris Bioscience™ Supplier No. 2303/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective α7 nAChR agonist

Highly selective α7 nAChR agonist (Ki = 26 nM) displaying negligible blockade of α1β1γδ and α3β4 nAChRs (IC50 ≥ 60 μM). Found to be inactive against a panel of 32 receptors at 1 μM, except 5-HT3 receptors (Ki = 930 nM).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 123464-89-1
Molecular Formula C14H20Cl2N2O2

Specifications

Quantity 10 mg
Formula Weight 264.75
Purity >98%
Molecular Weight (g/mol) 319.23
Chemical Name or Material PNU 282987
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.