Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PNU 74654
GREENER_CHOICESDP

Catalog No. 353425 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-342-5 25 mg
35-345 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 35-342-5 Supplier Tocris Bioscience™ Supplier No. 3534/25
Add to Cart
Add to Cart
This item is not returnable. View return policy

β-catenin binder; inhibits Wnt signaling

Binds to β-catenin (KD = 450 nM). Inhibits the interaction between β-catenin and T cell factor 4 (Tcf4) and disrupts the Wnt signaling pathway.

Specifications

Chemical Name or Material 2-Phenoxybenzoic acid-[(5-methyl-2-furanyl)methylene]hydrazide
CAS 113906-27-7
Assay Percent Range >99%
Quantity 25 mg
Molecular Formula C19H16N2O3
Formula Weight Observed MW: 320.34
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.