Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PNUTS/PPP1R10 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100604AF350
Description
PNUTS/PPP1R10 Polyclonal antibody specifically detects PNUTS/PPP1R10 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
PNUTS/PPP1R10 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
CAT53, FB19PP1-binding protein of 114 kDa, MHC class I region proline-rich protein CAT53, p99, Phosphatase 1 nuclear targeting subunit, phosphatase nuclear targeting subunit, PNUTSPP1R10, Protein FB19, protein phosphatase 1 regulatory subunit 10, protein phosphatase 1, regulatory (inhibitor) subunit 10, protein phosphatase 1, regulatory subunit 10, serine/threonine-protein phosphatase 1 regulatory subunit 10 | |
A synthetic peptide, mapping to a region region between residues 350 and 400 of human Phosphatase Nuclear Targeting Subunit (a.k.a. Protein Phosphatase 1, regulatory subunit 10) using the numbering given in TrEMBL entry Q96QC0 (GeneID 5514). | |
0.1 mL | |
Cellular Markers, Protein Phosphatase | |
5514 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction