Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PNUTS/PPP1R10 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP23880125UL
Description
PNUTS/PPP1R10 Polyclonal specifically detects PNUTS/PPP1R10 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
PNUTS/PPP1R10 | |
Polyclonal | |
Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Q96QC0 | |
PPP1R10 | |
This antibody was developed against a recombinant protein corresponding to amino acids: LMDTASLEPGALDAKPVESPGDPNQLTRKGRKRKSVTWPEEGKLREYFYFELDETERVNVNKIKDFGEAAKREILSDRHAFETARRLSHDNMEEKVPWV | |
25 μL | |
Cellular Markers, Protein Phosphatase | |
5514 | |
Human | |
IgG |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
CAT53, FB19PP1-binding protein of 114 kDa, MHC class I region proline-rich protein CAT53, p99, Phosphatase 1 nuclear targeting subunit, phosphatase nuclear targeting subunit, PNUTSPP1R10, Protein FB19, protein phosphatase 1 regulatory subunit 10, protein phosphatase 1, regulatory (inhibitor) subunit 10, protein phosphatase 1, regulatory subunit 10, serine/threonine-protein phosphatase 1 regulatory subunit 10 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction