Learn More
Roche Diagnostics POC Chemstrip Urine Test Strips
Catalog No. 22045621 Shop All Roche Diagnostics Products
Click to view available options
Detectable Analytes:
Bilirubin, Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Specific Gravity, Urobilinogen
Bilirubin, Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Urobilinogen
Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein
Blood, Glucose, Leukocyte, Nitrite, Protein
Glucose, Protein
Leukocyte, Nitrite
Type:
Chemstrip 10 MD
Chemstrip 10 SG
Chemstrip 2 GP
Chemstrip 2 LN
Chemstrip 5 OB
Chemstrip 7
Chemstrip 9
This item is not returnable. View return policy
Catalog No. 22-045-621
Supplier: Roche Diagnostics 11893467160
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Accuracy, performance and testing simplicity
  • Avoid potentially costly consequences of missed diagnoses — vitamin C interference is virtually eliminated on the glucose pad1, 2
  • Avoid false negatives — both acetoacetic acid and acetone are detected on the ketone reagent pad3
  • Simplify and streamline reading — all reagent pads are read at one time, between one and two minutes after dipping3
  • Promote safe and hygienic testing practice — fingers stay clear of specimen
  • Generate confidence in test result accuracy — Chemstrip construction allows uniform liquid penetration and prevents runover onto adjacent pads
TRUSTED_SUSTAINABILITY
Specifications
Clia Complexity Waived
Detectable Analytes Blood, Glucose, Leukocyte, Nitrite, Protein
Type Chemstrip 5 OB
For Use With (Application) Urinalysis
Quantity 100 Pack
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.